7-Aminoclonazepam (CRM), Cayman Chemical
76544-178
:
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:4959-17-5
- Description:7-Aminoclonazepam (CRM)
- Size:1 mg
- Synonym(s):Aminoclonazepam; Chloraminazepam
- Formula:C₁₅H₁₂ClN₃O
- Storage Temperature:–20 °C
- Description:7-Aminoclonazepam
- Cat. No.:76544-178
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard