para-Fluorobutyryl fentanyl-d7 (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-148
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- Description:para-Fluorobutyryl fentanyl-d7 (hydrochloride) (CRM)
- Size:100 µg
- Synonym(s):4-FBF-d7; p-FBF-d7; para-FBF-d7; 4-Fluorobutyryl fentanyl-d7; p-Fluorobutyryl fentanyl-d7
- Formula:C₂₃H₂₂D₇FN₂O·HCl
- Storage Temperature:–20 °C
- Description:para-Fluorobutyryl fentanyl-d7 (hydrochloride)
- Cat. No.:76544-148
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard