Order Entry
Puerto Rico
ContactUsLinkComponent
MAB-CHMINACA-d4 (CRM), Cayman Chemical
MAB-CHMINACA-d4 (CRM), Cayman Chemical
Certificates
MAB-CHMINACA-d4 (CRM), Cayman Chemical

Specifications

  • CAS No.:
    2747913-91-1
  • Synonym(s):
    ADB-CHMINACA-d4
  • Formula:
    C₂₁H₂₆D₄N₄O₂
  • Storage Temperature:
    –20 °C
  • Description:
    MAB-CHMINACA-d4

Specifications

Frequently Bought Together