Order Entry
Puerto Rico
ContactUsLinkComponent
Ocfentanil (hydrochloride) (CRM), Cayman Chemical
Ocfentanil (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-112
CAS Number:  
Ocfentanil (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-112
Supplier Number:  19635-100
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • CAS No.:
    112964-97-3
  • Description:
    Ocfentanil (hydrochloride) (CRM)
  • Size:
    100 µg
  • Synonym(s):
    A-3217; o-fluoro MAF; ortho-fluoro MAF; o-fluoro Methoxyacetyl fentanyl; ortho-fluoro Methoxyacetyl fentanyl
  • Formula:
    C₂₂H₂₇FN₂O₂·HCl
  • Storage Temperature:
    –20 °C
  • Description:
    Ocfentanil (hydrochloride)
  • Cat. No.:
    76544-112

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard