7-Aminonimetazepam (CRM), Cayman Chemical
Catalog # 76544-074
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:4959-16-4
- Description:7-Aminonimetazepam (CRM)
- Size:1 mg
- Synonym(s):Ro 05-3418; Ro 05-4318
- Formula:C₁₆H₁₅N₃O
- Storage Temperature:–20 °C
- Description:7-Aminonimetazepam
- Cat. No.:76544-074
Specifications
About this item
A metabolite of nimetazepam that can be detected in urine specimens following nimetazepam use
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard