Temazepam-d5 (CRM), Cayman Chemical
76544-004
:
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:136765-51-0
- Description:Temazepam-d5 (CRM)
- Size:1 mg
- Synonym(s):N-Methyloxazepam-d5
- Formula:C₁₆H₈ClD₅N₂O₂
- Storage Temperature:–20 °C
- Description:Temazepam-d5
- Cat. No.:76544-004
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard