5-fluoro PB-22 3-carboxyindole metabolite-d5 (CRM), Cayman Chemical
76543-972
:
- Concentration:100 µg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- Description:5-fluoro PB-22 3-carboxyindole metabolite-d5 (CRM)
- Size:100 µg
- Formula:C₁₄H₁₁D₅FNO₂
- Storage Temperature:–20 °C
- Description:5-fluoro PB-22 3-carboxyindole metabolite-d5
- Cat. No.:76543-972
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard