AB-CHMINACA metabolite M4-d4 (CRM), Cayman Chemical
76543-970
:
- Concentration:100 µg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- Description:AB-CHMINACA metabolite M4-d4 (CRM)
- Size:100 µg
- Formula:C₁₅H₁₄D₄N₂O₂
- Storage Temperature:–20 °C
- Description:AB-CHMINACA metabolite M4-d4
- Cat. No.:76543-970
An internal standard for the quantification of AB-CHMINACA metabolite M4 by GC- or LC-MS
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard