Cannabidiolic Acid (CRM), Cayman Chemical
Catalog # 76543-962
Supplier: Cayman Chemical Company
CSA Certified
CAS Number:
Specifications
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:1244-58-2
- Description:Cannabidiolic Acid (CRM)
- Size:1 mg
- Synonym(s):CBDA; β-Resorcylic Acid
- Formula:C₂₂H₃₀O₄
- Storage Temperature:–20 °C
- Description:Cannabidiolic acid
- Cat. No.:76543-962
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard