Masterflex® Multi-Syringe Pumps, Avantor®
Supplier: VWR International
Certificates
About this item
Economical & packed with features.
- Flow rates up to 147 ml/min per syringe
- Choose pumps which accept two or up to ten syringes—ideal for pumping exact proportions
- RS-232 interface lets you easily control pumps from your computer
- Backlit alphanumeric LCD steps you through setup
Simple keypad programming will have you up and running quickly. Program infusion/withdrawal pumps to sequence operations: either withdrawal followed by infusion or infusion followed by withdrawal.
Pumps feature two TTL inputs and two TTL outputs. The TTL
Multistep programming is available for these models. Please contact your local Avantor office for assistance.
- Environmentally Preferable: