Specifications
- Description:Connector with Wi-Fi
- Environmentally Preferable:
- Cat. No.:76542-102
Specifications
About this item
LIS connector base for use with Clinitek Status™ instrument.
- Connects by wireless, Ethernet or serial transmission
- Supports HL7 and POCT1-A2 protocols
- Integrates data into LIS/HIS/EMR or middleware data-management systems
- Interfaces to Siemens Healthineers POCcelerator™ system, UniPOC™ system, RAPIDCOMM® data management system, and third-party data-management software solutions
Offers flexible connectivity solutions by integrating data directly to the LIS, EMR or via point-of-care data management software solutions. Improves risk management through advanced operator control functions, prevents unauthorized use.
Proprietary Auto-Checks® technology, prevents false-positive results if test strip integrity is compromised by humidity overexposure and ensures that only validated Siemens Healthineers test strips are used to test patient samples.