Order Entry
Puerto Rico
ContactUsLinkComponent
 
Toxic
Health
Acrylamide : Bisacrylamide (29:1) 40% in aqueous solution
Catalog # 76461-626
Supplier:  MP Biomedicals
CAS Number:  
Toxic
Health
Acrylamide : Bisacrylamide (29:1) 40% in aqueous solution
Catalog # 76461-626
Supplier:  MP Biomedicals
Supplier Number:  11BIAC2902
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 L
  • Cat. No.:
    76461-626

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
 :79-06-1
MDL Number:MFCD00080848