ContactUsLinkComponent
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.2 µm, Whatman products (Cytiva)
 :  Cytiva
undefined
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.2 µm, Whatman products (Cytiva)
 :  Cytiva
  • Catalog No:
  • 89233-756
  • 10034-928
  • 10034-926
  • 10035-540
  • 89233-760
  • 89233-762
  • 89233-758
  • Max. Operating Temperature:
  • 130°C
  • 130°C
  • 130°C
  • 130°C
  • 130°C
  • 130°C
  • 130°C
  • Autoclavable:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Burst Strength:
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • Porosity:
  • 74% min. to 77% max.
  • 74% min. to 77% max.
  • 74% min. to 77% max.
  • 74% min. to 77% max.
  • 74% min. to 77% max.
  • 74% min. to 77% max.
  • 74% min. to 77% max.
  • Protein:
  • Medium Protein Binding
  • Medium Protein Binding
  • Medium Protein Binding
  • Medium Protein Binding
  • Medium Protein Binding
  • Medium Protein Binding
  • Medium Protein Binding
  • Solvent:
  • Medium Solvent Resistance
  • Medium Solvent Resistance
  • Medium Solvent Resistance
  • Medium Solvent Resistance
  • Medium Solvent Resistance
  • Medium Solvent Resistance
  • Medium Solvent Resistance