Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Thymolphthalein ACS

Thymolphthalein ACS

Thymolphthalein ACS
Certificates
Thymolphthalein ACS

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:12,09542
  • Melting Pt:251…253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Visual transition interval:
    from pH 8.8 (colorless) to pH 10.5 (blue)
  • Clarity of solution:
    passes test
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $39.90
    $39.90
    Each (10g)
       $39.90

    Supplier Number: 42095-0100
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $77.80
    $77.80
    Each (25g)
       $77.80

    Supplier Number: 42095-0250
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $185.00
    $185.00
    Each (100g)
       $185.00

    Supplier Number: 42095-1000
  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $799.00
    $799.00
    Each (500g)
       $799.00

    Supplier Number: 42095-5000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.