Vanillin 99%, pure
About this item
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Merck Index:13,09998
- Boiling Pt:170 °C (15 mmHg)
- Melting Pt:81…84 °C
- Flash Pt:147 °C
- Storage Temperature:Ambient
- EINECS:204-465-2
4 Options Available Below
Product Details & Documents
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Merck Index:13,09998
- Boiling Pt:170 °C (15 mmHg)
- Melting Pt:81…84 °C
- Flash Pt:147 °C
- Storage Temperature:Ambient
- EINECS:204-465-2
Specifications for Product Family
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:81 to 84 °C
- Titration with NaOH:≥98.5 %
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 14082-0020
- Supplier Number: 14082-1000
- Plastic bottle250 gDiscontinued-Supplier Number: 14082-2500
- Supplier Number: 14082-0010
- Supplier Number: 14082-0050
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



