



Hydrazine hydrate 100% (∼64% w/w N₂H₄)
Catalog # TS19671-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle for solids
- Size:100 g
- Cat. No.:TS19671-1000
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Refractive index:1.4285 to 1.4315 (20°C, 589 nm)
- Density:(20°C) 1.030 to 1.033 g/ml
- Titration with HCl:63.6 to 64.6 % (N2H4)
Specification Test Results
About this item
:
:32.05 g/mol (anhy)
Merck Index:1304790
Boiling Pt:117.2 °C (1013 hPa)
Melting Pt:–51.5 °C
Density:1.0315 g/cm³
Flash Pt:75 °C
EINECS:206-114-9
UN:2030
ADR:8,II
:GHS06
:GHS08
:GHS05
:GHS09
CAS Number:10217-52-4
:7803-57-8
Formula:NH₂NH₂·xH₂O
MDL Number:MFCD00149931