
Sudan II, pure
Catalog # TS19015-0250
Supplier: Thermo Scientific Chemicals
CAS Number:
Specification Test Results
- Appearance:Red to orange-brown Powder
- Infrared spectrum:Conforms
- UV Lambda max:at approximately 420 and 493 nm (in methanol)
- Specific extinction E (1%/1cm):≥500 (at 493 nm, in methanol)