Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lithium acetylide ethylenediamine complex 85%

Lithium acetylide ethylenediamine complex 85%

Irritant
Corrosive
Flammable
Lithium acetylide ethylenediamine complex 85%
Certificates
Lithium acetylide ethylenediamine complex 85%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
  • CAS Number:6867-30-7
  • Formula:C₄H₉LiN₂
  • MDL Number:MFCD00013183
  • Molecular Weight:92.07 g/mol
  • Boiling Pt:110.6 °C
  • Melting Pt:76 °C
  • Storage Temperature:Refrigerator
  • UN:3131
  • ADR:4.3,I
Specifications for Product Family

Specification Test Results

  • Appearance:
    Light yellow to light brown Crystalline powder or crystals
  • Assay:
    ≥80 % (Gasvolumetric)
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Metal can
    50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $167.00
    $167.00
    Each (50g)
       $167.00

    Supplier Number: 18125-0500
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Metal can
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $274.00
    $274.00
    Each (100g)
       $274.00

    Supplier Number: 18125-1000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.