

Methyl yellow, pure indicator grade
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:60-11-7
:
Formula:C₁₄H₁₅N₃
MDL Number:MFCD00008308
:225.29 g/mol
Merck Index:1203279
Melting Pt:111…117 °C
Storage Temperature:Ambient
EINECS:200-455-7
UN:3143
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Orange to orange-brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤3 %
- Appearance of solution:(0.1% soln. in ethanol) clear
- Visual transition interval:from pH 2.9 (red) to pH 4.0 (yellow)