

4-Picolyl chloride hydrochloride 97%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:1822-51-1
:
Formula:C₆H₇Cl₂N
MDL Number:MFCD00012826
Melting Pt:160…163 °C
Storage Temperature:Ambient
EINECS:217-350-7
UN:3261
ADR:8,III
:164.03 g/mol
:GHS07
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥96.0 %
- GC:≥99.4 %
- Color scale:≤20 APHA
Specification Test Results
Frequently Bought Together