Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Hydroquinone 99%

Irritant
Health
Corrosive
Environment
Hydroquinone 99%
Certificates
Hydroquinone 99%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  1,4-Dihydroxybenzene, Quinol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  • Merck Index:1304833
  • Boiling Pt:285…287 °C (760 mmHg)
  • Melting Pt:170…174 °C
  • Flash Pt:165 °C
  • Storage Temperature:Ambient
  • EINECS:204-617-8
  • UN:3077
  • ADR:9,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    170 to 175 °C
  • HPLC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $55.80
    $55.80
    Each (500g)
       $55.80

    Supplier Number: 12091-5000
  • Plastic bottle
    2 kg
    Log in for availability
    Log in for availability
    Each (2kg)
    Each (2kg) - $153.00
    $153.00
    Each (2kg)
       $153.00

    Supplier Number: 12091-0020
  • Plastic drum
    10 kg
    Log in for availability
    Log in for availability
    Each (10kg)
    Each (10kg) - $622.00
    $622.00
    Each (10kg)
       $622.00

    Supplier Number: 12091-0100

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.