3-Hydroxytyrosol
About this item
- CAS Number:10597-60-1
- Formula:C₈H₁₀O₃
- MDL Number:MFCD01320529
- Molecular Weight:154.17 g/mol
- Storage Temperature:Freezer
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
- CAS Number:10597-60-1
- Formula:C₈H₁₀O₃
- MDL Number:MFCD01320529
- Molecular Weight:154.17 g/mol
- Storage Temperature:Freezer
Specifications for Product Family
Specification Test Results
- Appearance:Liquid, Clear colorles to yellow
- Infrared spectrum:Conforms
- Refractive index:1.5810 to 1.5840 (20°C, 589 nm)
- HPLC:>=97.5 %
- Water:=<5 %
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: 46763-0010Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: 46763-0050Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



