Order Entry
Puerto Rico
ContactUsLinkComponent
Phenazopyridine Hydrochloride, 99%, Analytical Standard
Phenazopyridine Hydrochloride, 99%, Analytical Standard
Catalog # TS44192-1000
CAS Number:  
Phenazopyridine Hydrochloride, 99%, Analytical Standard
Catalog # TS44192-1000
Supplier Number:  44192-1000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • CAS No.:
    136-40-3
  • Description:
    Phenazopyridine hydrochloride, 99%
  • Size:
    100 mg
  • Formula:
    C₁₁H₁₁N₅.HCl
  • Description:
    Phenazopyridine hydrochloride
  • Cat. No.:
    TS44192-1000

Specifications

About this item