Electrode Storage Solution, Thermo Scientific
TS61121-0010
:
- Size:1 L
- Density:1
- Boiling Point:100°C (760mmHg)
- Cat. No.:TS61121-0010
CAS No.:
MDL No.: MFCD00211908
Density: 1
Boiling Point: 100 °C (760 mm Hg)
Appearance: Clear colorless Liquid
Storage Temperature: Ambient