Order Entry
Puerto Rico
ContactUsLinkComponent
 
4-Epioxytetracycline, 97%, can be used as secondary standard
  TS25771-1000
 :  
undefined
4-Epioxytetracycline, 97%, can be used as secondary standard
  TS25771-1000
 :  25771-1000
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pack type:
    Glass bottle
  • CAS No.:
    14206-58-7
  • Description:
    14206-58-7
  • Size:
    100 mg
  • Formula:
    C₂₂H₂₄N₂O₉
  • Description:
    4-Epioxytetracycline
  • Cat. No.:
    TS25771-1000

 

 

CAS No.: 14206-58-7

MDL No.: MFCD00021181
GHS Symbols: GHS07, GHS08,
Melting Point: 168 °C (D)
Appearance: Green-beige to brown Powder
Storage Temperature: Ambient
Signal word: Warning