Order Entry
Puerto Rico
ContactUsLinkComponent
Ascarite(II)® CO₂ Absorbent, 8 to 20 Mesh, Product of Arthur H. Thomas Company, Thermo Scientific
Ascarite(II)® CO₂ Absorbent, 8 to 20 Mesh, Product of Arthur H. Thomas Company, Thermo Scientific
Catalog # TS20808-5000
undefined
Ascarite(II)® CO₂ Absorbent, 8 to 20 Mesh, Product of Arthur H. Thomas Company, Thermo Scientific
Catalog # TS20808-5000
Supplier Number:  20808-5000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 g
  • Cat. No.:
    TS20808-5000
  • Packaging:
    Glass bottle

Specifications

About this item

CAS No.: 81133-20-2

MDL No.: MFCD00132264
UN: 1823
Hazard Class: 8
Packing group: II
GHS Symbols: GHS05
Appearance: Brown Granules
Storage Temperature: Ambient
Signal word: Danger