Ethanol absolute, anhydrous ≥99.5% (200 Proof)
:64-17-5
:
:C₂H₅OH
:MFCD00003568
:46.07 g/mol
:1170
:3,II
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3585 to 1.3605 (25 °C, 589 nm)
- Assay:≥99.5 % v/v
- Water:≤0.05 % (K.F.)
- Solubility:(in water) Passes test
- Color scale:≤10 APHA
- Specific gravity:(15.56°C) ≤0.7964
- Residue after evaporation:≤0.0025 %
- Substance darkened by H2SO4:Passes test
- Titrable acid:≤0.0005 meq/g
- Titrable base:≤0.0002 meq/g
- KMnO4-reducing substance:Passes test