Order Entry
Puerto Rico
ContactUsLinkComponent
Ethanol denatured for HPLC (denatured with 5% isopropanol and 5% methanol)
Ethanol denatured for HPLC (denatured with 5% isopropanol and 5% methanol)
Catalog # TS61105-0040
CAS Number:  
undefined
Ethanol denatured for HPLC (denatured with 5% isopropanol and 5% methanol)
Catalog # TS61105-0040
Supplier Number:  61105-0040
CAS Number:  

Specifications

  • Size:
    4 L
  • Cat. No.:
    TS61105-0040

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Refractive index:
    1.3580 to 1.3610 (25 °C, 589 nm)
  • Assay:
    89 to 91 %
  • Water:
    ≤0.10 %
  • Additional info:
    isopropyl alcohol: 4.5 - 5.5%
  • Solubility:
    (in water) Passes test
  • UV:
    at 205 nm A: ≤1.00
  • Color scale:
    ≤5 APHA
  • Identification:
    passes test
  • Residue after evaporation:
    ≤5 ppm
  • Titrable acid:
    ≤0.0003 meq/g
  • Titrable base:
    ≤0.0002 meq/g
  • Methanol:
    4.0 to 5.0 %
  • KMnO4-reducing substance:
    Passes test

Specification Test Results

About this item

 :
 :46.07 g/mol
UN:1170
ADR:3,III
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568