Silicon dioxide
Plastic bottle, 150 g
Catalog # TS40373-1500
Supplier: Thermo Scientific Chemicals
Supplier #: 40373-1500
Jump to:
Product Details & Documents
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- CAS Number:7631-86-9
- Formula:SiO₂
- MDL Number:MFCD00011232
- Molecular Weight:60.08 g/mol
- Boiling Pt:2230 °C (1013 hPa)
- Melting Pt:1719 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White Powder
- Density:≤60 g/l tamped density
- Particle size:≤0.02 % 325 Mesh residue
- Surface area:175 to 225 m²/g (BET)
- pH:3.8 to 4.3 (4 % slurry in water)
- Loss on heating:≤1.5 % (105°C, 2 h)
Specifications
- Pack type:Plastic bottle
- Size:150 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



