Silica gel 2-5 mm, non toxic for drying, with moisture indicator (orange gel)
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange Beads
- Bead size:2 to 5 mm
- Color:Orange (dry) to Colorless (wet)
- Residual water:≤2 % (150°C)
- Water adsorption:≥20 % (50% rH, 1 bar, 25°C)
Specification Test Results
Frequently Bought Together