Order Entry
Puerto Rico
ContactUsLinkComponent
Silica gel, Pore size: 60 Å 40-60 µm, Ultrapure for column chromatography
Silica gel, Pore size: 60 Å 40-60 µm, Ultrapure for column chromatography
Catalog # TS36005-0010
CAS Number:  
undefined
Silica gel, Pore size: 60 Å 40-60 µm, Ultrapure for column chromatography
Catalog # TS36005-0010
Supplier Number:  36005-0010
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg
  • Cat. No.:
    TS36005-0010

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Particle size:
    ≤10 % (Undersize)
  • Specific surface area:
    460 to 510 m²/g
  • Pore volume:
    0.75 to 0.80 ml/g
  • pH:
    6.5 to 7.0 (10 % suspension in water)
  • Volatile matter:
    ≤6.0 % (160°C)

Specification Test Results

About this item

 :
 :60.08 g/mol
Storage Temperature:Ambient
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232