Silica gel, Pore size: 60 Å 0.060-0.200 mm for chromatography
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Particle size:0.060 to 0.200 mm
- Specific gravity:0.46 to 0.55 kg/l
- pH:6.5 to 7.5 (5 % suspension in water)
- Residual water:2 to 4 % (160°C)
Specification Test Results
Frequently Bought Together