Silica gel, Pore size: 60 Å 0.035-0.070 mm for chromatography
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Particle size:0.035 to 0.070 mm
- Specific gravity:0.46 to 0.55 kg/l (Typical)
- pH:6.5 to 7.5 (5 % suspension in water) Typical
- Residual water:≤6 % (160°C, 15 min)
Specification Test Results
Frequently Bought Together