Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Polysorbate 20

Polysorbate 20

Polysorbate 20
Certificates
Polysorbate 20

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C (760 mmHg)
  • Density:1.1 g/cm³
  • Flash Pt:>150 °C
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear to slightly hazy yellow to brown-yellow Viscous liquid (May develop some turbidity or precipitate)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4680 to 1.4720 (20°C, 589 nm)
  • Water:
    ≤3 %
  • Dioxane:
    ≤10 ppm
  • Acid value:
    ≤2 mg KOH/g
  • Hydroxyl value:
    96 to 108 mg KOH/g
  • Saponification value:
    40 to 50 mg KOH/g
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    250 ml
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $52.60
    $52.60
    Each (250ml)
       $52.60

    Supplier Number: 23336-2500
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $97.10
    $97.10
    Each (1l)
       $97.10

    Supplier Number: 23336-0010
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $285.00
    $285.00
    Each (5l)
       $285.00

    Supplier Number: 23336-0051

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.