

Hydrochloric acid 1 M in ethyl acetate
TS38870-1000
:
- Pack type:Plastic bottle
- Size:100 ml
- Cat. No.:TS38870-1000
- Appearance:Clear colorless Solution
- Strength:0.95 to 1.15 M
:
:110 °C (1013 hPa)
:–30 °C
:0.9 g/cm³
:–3 °C
:Ambient
:2924
:3,II
:36.46
:GHS05
:GHS02
:7647-01-0
:HCl
:MFCD00011324