Order Entry
Puerto Rico
ContactUsLinkComponent
Bromine 99+%, extra pure
Toxic
Corrosive
Environment
Bromine 99+%, extra pure
Catalog # TS40284-5000
CAS Number:  
undefined
Toxic
Corrosive
Environment
Bromine 99+%, extra pure
Catalog # TS40284-5000
Supplier Number:  40284-5000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, plastic-coated
  • Size:
    500 ml
  • Cat. No.:
    TS40284-5000

Specifications

Specification Test Results

  • Appearance:
    Red-brown Liquid
  • Titration Iodometric:
    ≥99.0 %
  • Chlorine (Cl):
    ≤500 ppm
  • Iodine (I):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤2 ppm

Specification Test Results

About this item

 :
 :159.81 g/mol
Merck Index:13,01378
Boiling Pt:58.7 °C (1013 hPa)
Melting Pt:–7.2 °C
Density:3.111 g/cm³
Storage Temperature:Ambient
EINECS:231-778-1
UN:1744
ADR:8,I
 :GHS06
 :GHS05
 :GHS09
CAS Number:7726-95-6
Formula:Br₂
MDL Number:MFCD00010896