Vinylene carbonate 98% stabilised
Supplier: Thermo Scientific Chemicals
About this item
Synonyms: 1,3-Dioxol-2-one
- CAS Number:872-36-6
- Formula:C₃H₂O₃
- MDL Number:MFCD00005380
- Molecular Weight:86.05 g/mol
- Boiling Pt:162 °C (760 mmHg)
- Melting Pt:19…22 °C
- Density:1.36 g/cm³
- Flash Pt:73 °C
- Storage Temperature:Refrigerator
- UN:2810
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Synonyms: 1,3-Dioxol-2-one
- CAS Number:872-36-6
- Formula:C₃H₂O₃
- MDL Number:MFCD00005380
- Molecular Weight:86.05 g/mol
- Boiling Pt:162 °C (760 mmHg)
- Melting Pt:19…22 °C
- Density:1.36 g/cm³
- Flash Pt:73 °C
- Storage Temperature:Refrigerator
- UN:2810
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid or solid
- Infrared spectrum:Conforms
- Refractive index:1.4200 to 1.4230 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:≤0.1 % BHT
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 14089-0050Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: 14089-0250Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



