Order Entry
Puerto Rico
ContactUsLinkComponent
2-Dimethylaminoethyl methacrylate 99% stabilized
Irritant
2-Dimethylaminoethyl methacrylate 99% stabilized
Catalog # TS21584-0010
CAS Number:  
undefined
Irritant
2-Dimethylaminoethyl methacrylate 99% stabilized
Catalog # TS21584-0010
Supplier Number:  21584-0010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 kg
  • Cat. No.:
    TS21584-0010

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4410 (20°C, 589 nm)
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥98.5 %
  • Stabilizer:
    750 to 2000 ppm MEHQ

Specification Test Results

About this item

 :
 :157.21 g/mol
Melting Pt:–50 °C
Flash Pt:65 °C
Storage Temperature:Refrigerator
UN:2522
ADR:6.1,II
 :GHS07
CAS Number:2867-47-2
Formula:C₈H₁₅NO₂
MDL Number:MFCD00008589