


Giemsa's stain solution biological stain
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:51811-82-6
:
MDL Number:MFCD00081642
Density:0.99 g/cm³
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:257-438-2
UN:1230
ADR:3,II
:GHS02
:GHS06
:GHS08
Specification Test Results
- Appearance:Dark blue to dark purple Solution
- Additional info:Giemsa stain powder in MeOH, Glycerol mixture
- UV Lambda max:523 to 526 nm (c=0.1, MeOH) Eosine