



Tetrabutylammonium hydroxide 1 M in methanol
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:2052-49-5
:
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425
:259.48
Melting Pt:–98 °C
Density:0.83 g/cm³
Flash Pt:12 °C
EINECS:218-147-6
UN:3286
ADR:3,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Solution
- Titration with HCl:0.95 to 1.10 M