Bromothymol blue ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:76-59-5
:
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872
Merck Index:13,01430
Melting Pt:204 °C (dec.)
:624.39 g/mol
Storage Temperature:Ambient
EINECS:200-971-2
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Light pink to brown to purple Crystalline powder
- Infrared spectrum:Conforms
- Visual transition interval:from pH 6.0 (yellow) to pH 7.6 (blue)
- Clarity of solution:Passes test
- Specific optical rotation:+103° to +108° (20°C, 589 nm) (c=5, H2O)