ContactUsLinkComponent
Ethylene glycol monomethyl ether (2-methoxyethanol) for HPLC
Irritant
Health
Flammable
Ethylene glycol monomethyl ether (2-methoxyethanol) for HPLC
Certificates
undefined
Irritant
Health
Flammable
Ethylene glycol monomethyl ether (2-methoxyethanol) for HPLC
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.0 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Residue after evaporation:
    ≤0.001 %
Specification Test Results