Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ethanolamine 99%

Ethanolamine 99%

Glass bottle, 1 L
Catalog # TS14958-0010
Irritant
Corrosive
Supplier #:  14958-0010
Ethanolamine 99%
Catalog # TS14958-0010
CAS Number:  141-43-5
Ethanolamine 99%
Ethanolamine 99%

Glass bottle, 1 L

CAS Number:  141-43-5
Price
$77.20
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Boiling Pt:171 °C (1013 hPa)
  • Melting Pt:10.3 °C
  • Density:1.012 g/cm³
  • Flash Pt:92 °C
  • Storage Temperature:Ambient
  • UN:2491
  • ADR:8,III
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4530 to 1.4550 (20°C, 589 nm)
  • Water:
    ≤0.5 %
  • GC:
    ≥98.5 %
  • Color scale:
    ≤15 APHA

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.