Order Entry
Puerto Rico
ContactUsLinkComponent
Calcium sulfate, Drierite™ 8 mesh for drying, with indicator
Calcium sulfate, Drierite™ 8 mesh for drying, with indicator
Certificates
undefined
Calcium sulfate, Drierite™ 8 mesh for drying, with indicator

Specification Test Results

  • Appearance:
    Blue Irregular granules
  • Water:
    ≤0.3 %
  • Particle size:
    8 mesh
  • Composition:
    ≥98 % (CaSO4) Typical

Specification Test Results