

N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:127-19-5
:
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:164…166 °C (760 mmHg)
Melting Pt:–20 °C
Density:0.937 g/cm³
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.05 %
- GC:≥99.80 %
- UV:1 cm cell vs HPLC-grade water
- Non-volatile matter:≤0.0003 %
- Acidity:≤0.0005 meq/g
- Alkalinity:≤0.003 meq/g