Order Entry
Puerto Rico
ContactUsLinkComponent
 
di-Potassium hydrogen phosphate, anhydrous 99% for HPLC
Catalog # TS44736-2500
CAS Number:  
undefined
di-Potassium hydrogen phosphate, anhydrous 99% for HPLC
Catalog # TS44736-2500
Supplier Number:  44736-2500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g
  • Cat. No.:
    TS44736-2500

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals or granules
  • Loss on drying:
    ≤1 %
  • Solubility:
    Clear colorless
  • UV:
    (10 % in water)
  • pH:
    8.5 to 9.6 (10 % in water)
  • Titration with HClO4:
    ≥98.5 %

Specification Test Results

About this item

 :
 :174.18 g/mol
Melting Pt:340 °C
Storage Temperature:Ambient
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383