Order Entry
Puerto Rico
ContactUsLinkComponent
1,2-Dimethoxyethane, extra dry 99+% stabilized, AcroSeal™
Irritant
Health
Flammable
1,2-Dimethoxyethane, extra dry 99+% stabilized, AcroSeal™
Catalog # TS61035-0010
CAS Number:  
undefined
Irritant
Health
Flammable
1,2-Dimethoxyethane, extra dry 99+% stabilized, AcroSeal™
Catalog # TS61035-0010
Supplier Number:  61035-0010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    TS61035-0010

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.0 %
  • Stabilizer:
    Approx. 100 ppm BHT
  • Residue after ignition:
    ≤0.001 %

Specification Test Results

About this item

 :
 :90.12 g/mol
Merck Index:13,03251
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–69 °C
Density:0.867 g/cm³
Flash Pt:–6 °C
EINECS:203-794-9
UN:2252
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:110-71-4
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502