Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
2-Phenoxyethanol 99%

2-Phenoxyethanol 99%

Irritant
2-Phenoxyethanol 99%
Certificates
2-Phenoxyethanol 99%

5 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17 g/mol
  • Merck Index:12,07410
  • Boiling Pt:237 °C (760 mmHg)
  • Melting Pt:11…13 °C
  • Density:1.1 g/cm³
  • Flash Pt:130 °C
  • Storage Temperature:Ambient
  • EINECS:204-589-7
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5360 to 1.5400 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • pH:
    9.5 to 11.0 (5% at 25°C)
  • Titration with HClO4:
    ≥99 %
  • Iron (Fe):
    ≤0.001 %
Product Family Options

Product Information

  • Size
    Pack type
    Availability
    Price
  • 250 ml
    Glass bottle
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $29.90
    $29.90
    Each (250ml)
       $29.90

    Supplier Number: 13024-2500
  • 1 L
    Glass bottle
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $65.30
    $65.30
    Each (1l)
       $65.30

    Supplier Number: 13024-0010
  • 2.5 L
    Glass bottle
    Log in for availability
    Log in for availability
    Each (2,500ml)
    Each (2,500ml) - $128.00
    $128.00
    Each (2,500ml)
       $128.00

    Supplier Number: 13024-0025
  • 5 L
    Plastic drum
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $268.00
    $268.00
    Each (5l)
       $268.00

    Supplier Number: 13024-0051
  • 25 L
    Metal drum
    Log in for availability
    Log in for availability
    Each (25l)
    Each (25l) - $808.00
    $808.00
    Each (25l)
       $808.00

    Supplier Number: 13024-0250

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.