Dimethyl sulfoxide-[D₆] 99.9 atom % D for NMR spectroscopy
Catalog # TS32077-0075
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Size:10 x 0.75 ml
- Cat. No.:TS32077-0075
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.03 % (Coulometric)
- D-Enrichment:≥99.85 %
Specification Test Results
About this item
:
:84.09 g/mol
Boiling Pt:189 °C (5 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³
Flash Pt:88 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-617-0
CAS Number:2206-27-1
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090