Dimethyl sulfoxide 99.5+% for analysis
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (760 mmHg)
Melting Pt:18.4 °C
Density:1.1 g/cm³
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 Solvent selection guide recommended by default
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.50 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0002 meq/g
- Substance darkened by H2SO4:Passes test
- Iron (Fe):≤1 ppm